Learn More
Sigma Aldrich Fine Chemicals Biosciences D-(+)-Galactose, 59-23-4, MFCD00151230, 100 G
Catalog No. NC1391554
Encompass
Empirical Formula (Hill Notation): C6H12O6, Molecular Weight: 180.16, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture, Synonym: Galactose. D-(+)-Galactose has been used in cell line and culture medium to permits the cell growth arrest without altering the specific production rate of rh-tPA (qtPA).
Catalog No. NC1391554 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G5388100G
Empirical Formula (Hill Notation): C6H12O6, Molecular Weight: 180.16, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture, Synonym: Galactose. D-(+)-Galactose has been used in cell line and culture medium to permits the cell growth arrest without altering the specific production rate of rh-tPA (qtPA).