Learn More
Medchemexpress LLC β-D-Glucan | 9041-22-9 | 99.0% | 25 MG
SDP
Catalog No. 5000349192
Encompass_Preferred
β-D-Glucan is a natural, non-digestible polysaccharide known for its high biocompatibility. It is selectively recognized by receptors such as Dectin-1 and Toll-like receptors, and is readily internalized by murine or human macrophages, indicating its potential for targeted delivery applications. Additionally, it can function as an enteric delivery vehicle for probiotics.
  • Natural, non-digestible polysaccharide
  • High biocompatibility
  • Selectively recognized by Dectin-1 and Toll-like receptors
  • Easily internalized by murine or human macrophages
  • Potential for targeted delivery
  • Functions as an enteric delivery vehicle for probiotics
Catalog No. 50-003-49192 Supplier Medchemexpress LLC Supplier No. 25MGHY139413
β-D-Glucan is a natural, non-digestible polysaccharide known for its high biocompatibility. It is selectively recognized by receptors such as Dectin-1 and Toll-like receptors, and is readily internalized by murine or human macrophages, indicating its potential for targeted delivery applications. Additionally, it can function as an enteric delivery vehicle for probiotics.
  • Natural, non-digestible polysaccharide
  • High biocompatibility
  • Selectively recognized by Dectin-1 and Toll-like receptors
  • Easily internalized by murine or human macrophages
  • Potential for targeted delivery
  • Functions as an enteric delivery vehicle for probiotics