Learn More
Medchemexpress LLC β-D-glucan | 9041-22-9 | MFCD00466918 | 99.0% | 504.4 g/mol | C18H32O16 | 5 MG
SDP
Catalog No. 5000223226
Encompass_Preferred
β-D-Glucan is a natural, non-digestible polysaccharide with high biocompatibility used in research for immune recognition and delivery applications. It is recognized by immune receptors and is readily internalized by macrophages, supporting use in targeted delivery and as an enteric carrier for probiotics.
  • High purity (99.0%) for research applications.
  • Recognized by immune receptors such as Dectin-1 and toll-like receptors.
  • Efficient uptake by murine and human macrophages.
  • Suitable as an enteric delivery vehicle for probiotics.
  • Reported in vitro inhibition of breast cancer cell proliferation.
  • Available in small sample sizes for assay development.
Catalog No. 50-002-23226 Supplier Medchemexpress LLC Supplier No. HY1394135MG
β-D-Glucan is a natural, non-digestible polysaccharide with high biocompatibility used in research for immune recognition and delivery applications. It is recognized by immune receptors and is readily internalized by macrophages, supporting use in targeted delivery and as an enteric carrier for probiotics.
  • High purity (99.0%) for research applications.
  • Recognized by immune receptors such as Dectin-1 and toll-like receptors.
  • Efficient uptake by murine and human macrophages.
  • Suitable as an enteric delivery vehicle for probiotics.
  • Reported in vitro inhibition of breast cancer cell proliferation.
  • Available in small sample sizes for assay development.