Learn More
Medchemexpress LLC D-Glucaric acid | 87-73-0 | MFCD09752094 | 98.0% | 210.14 g/mol | C6H10O8 | 50 MG
SDP
Catalog No. 5000282561
Encompass_Preferred
Saccharic acid (D-glucaric acid) is a research-grade small molecule that acts as a competitive inhibitor of β-glucuronidase. Supplied as a white to off-white solid, it is used in biochemical and enzymology studies involving glucuronide metabolism and enzymatic inhibition assays.
  • High purity suitable for research applications.
  • Water soluble with high solubility in H2O (250 mg/mL); ultrasonic assistance recommended.
  • Defined chemical properties: C6H10O8, 210.14 g/mol.
  • Stable when stored as a powder at low temperatures for extended periods.
  • Available in small pack sizes appropriate for screening and assay development.
Catalog No. 50-002-82561 Supplier Medchemexpress LLC Supplier No. HYW34588550MG
Add to Cart
Add to Cart
Saccharic acid (D-glucaric acid) is a research-grade small molecule that acts as a competitive inhibitor of β-glucuronidase. Supplied as a white to off-white solid, it is used in biochemical and enzymology studies involving glucuronide metabolism and enzymatic inhibition assays.
  • High purity suitable for research applications.
  • Water soluble with high solubility in H2O (250 mg/mL); ultrasonic assistance recommended.
  • Defined chemical properties: C6H10O8, 210.14 g/mol.
  • Stable when stored as a powder at low temperatures for extended periods.
  • Available in small pack sizes appropriate for screening and assay development.