Learn More
Medchemexpress LLC D-Glucaric acid potassium | 576-42-1 | 98.0% | 250 MG
SDP
Catalog No. 5000343024
Encompass_Preferred
D-Glucaric acid potassium is an orally active end product of the D-glucuronic acid pathway found in mammals, also present in various fruits and vegetables. This compound demonstrates cholesterol-lowering and anti-tumor activities, with the ability to inhibit tumor cell proliferation and inflammation, and induce apoptosis.
  • Orally active end product of the D-glucuronic acid pathway
  • Found in various fruits and vegetables
  • Exhibits cholesterol-lowering activities
  • Exhibits anti-tumor activities
  • Inhibits tumor cell proliferation and inflammation
  • Induces apoptosis
Catalog No. 50-003-43024 Supplier Medchemexpress LLC Supplier No. 250MGHY128749
D-Glucaric acid potassium is an orally active end product of the D-glucuronic acid pathway found in mammals, also present in various fruits and vegetables. This compound demonstrates cholesterol-lowering and anti-tumor activities, with the ability to inhibit tumor cell proliferation and inflammation, and induce apoptosis.
  • Orally active end product of the D-glucuronic acid pathway
  • Found in various fruits and vegetables
  • Exhibits cholesterol-lowering activities
  • Exhibits anti-tumor activities
  • Inhibits tumor cell proliferation and inflammation
  • Induces apoptosis