Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucitol, 1,4:3,6-dianhydro-, 2,5-dinitrate | 87-33-2 | 99.89% | 25 MG
SDP

Catalog No. 5000414167
Encompass_Preferred

Isosorbide dinitrate (ISDN) is a Nitric Oxide (NO) donor that helps prevent left ventricular (LV) remodeling and degradation of cardiac function following myocardial infarction (MI).

  • Synonyms: ISDN
  • CAS No.: 87-33-2
  • Purity: 99.89%
  • Molecular weight: 236.14
  • Formula: C6H8N2O8
  • Appearance: Solid
  • Color: White to off-white
  • Handling: For research use only. Not sold to patients.
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).
  • Solubility: In DMSO: 100 mg/mL (423.48 mM); requires ultrasonic and newly opened hygroscopic DMSO.

Catalog No. 50-004-14167 Supplier Medchemexpress LLC Supplier No. 25MGHYB1409
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isosorbide dinitrate (ISDN) is a Nitric Oxide (NO) donor that helps prevent left ventricular (LV) remodeling and degradation of cardiac function following myocardial infarction (MI).

  • Synonyms: ISDN
  • CAS No.: 87-33-2
  • Purity: 99.89%
  • Molecular weight: 236.14
  • Formula: C6H8N2O8
  • Appearance: Solid
  • Color: White to off-white
  • Handling: For research use only. Not sold to patients.
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).
  • Solubility: In DMSO: 100 mg/mL (423.48 mM); requires ultrasonic and newly opened hygroscopic DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.