Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glucitol, 2,5-anhydro-, 1,6-bis(dihydrogen phosphate) | 4429-47-4 | MFCD01863342 | 99.5% | 324.12 | C6H14O11P2 | 5 MG
SDP

Catalog No. 5000221822
Encompass_Preferred

2,5-Anhydro-D-glucitol-1,6-diphosphate is a research-grade biochemical reagent that acts as a limited stimulator of yeast pyruvate kinase and serves as an analogue of 2,5-anhydro-D-mannitol 1,6-bisphosphate. It is supplied as a colorless to light-yellow oil of high purity for enzymology and biochemical assays. This product is for research use only.

  • Molecular formula C6H14O11P2; molecular weight 324.12
  • CAS number 4429-47-4
  • Purity 99.49%
  • Appearance colorless to light yellow oil
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (6 months), -20°C (1 month)
  • Solubility details available on product datasheet
  • Available in 5 mg pack
  • Intended for research use only

Catalog No. 50-002-21822 Supplier Medchemexpress LLC Supplier No. HY1341775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,5-Anhydro-D-glucitol-1,6-diphosphate is a research-grade biochemical reagent that acts as a limited stimulator of yeast pyruvate kinase and serves as an analogue of 2,5-anhydro-D-mannitol 1,6-bisphosphate. It is supplied as a colorless to light-yellow oil of high purity for enzymology and biochemical assays. This product is for research use only.

  • Molecular formula C6H14O11P2; molecular weight 324.12
  • CAS number 4429-47-4
  • Purity 99.49%
  • Appearance colorless to light yellow oil
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (6 months), -20°C (1 month)
  • Solubility details available on product datasheet
  • Available in 5 mg pack
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.