Learn More
Medchemexpress LLC D-Glucitol, 4-O-β-D-galactopyranosyl- | 585-86-4 | 100 MG
SDP
Catalog No. 5000418108
Encompass_Preferred
Lactitol (D-Lactitol) is a non-absorbable disaccharide primarily used for research purposes. It modulates gut flora by reducing harmful bacteria and increasing beneficial ones, making it suitable for studies on portosystemic encephalopathy and chronic constipation.
  • Non-absorbable disaccharide
  • Reduces populations of Bacteroides, Clostridium, coliforms, and Eubacterium
  • Increases populations of Lactobacillus and Streptococcus
  • Useful in research for portosystemic encephalopathy
  • Useful in research for chronic constipation
  • For research use only
Catalog No. 50-004-18108 Supplier Medchemexpress LLC Supplier No. HYN7104100MG
Lactitol (D-Lactitol) is a non-absorbable disaccharide primarily used for research purposes. It modulates gut flora by reducing harmful bacteria and increasing beneficial ones, making it suitable for studies on portosystemic encephalopathy and chronic constipation.
  • Non-absorbable disaccharide
  • Reduces populations of Bacteroides, Clostridium, coliforms, and Eubacterium
  • Increases populations of Lactobacillus and Streptococcus
  • Useful in research for portosystemic encephalopathy
  • Useful in research for chronic constipation
  • For research use only