Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Gluconic acid, 2-amino-2-deoxy- | 3646-68-2 | 98.0% | 250 MG
SDP

Catalog No. 5000341790
Encompass_Preferred

D-Glucosamic acid is an endogenous metabolite with a molecular weight of 195.17 and a chemical formula of C6H13NO6. It is a solid, typically appearing white to yellow, and is primarily used for research applications.

  • Endogenous metabolite
  • Molecular formula: C6H13NO6
  • Molecular weight: 195.17
  • CAS number: 3646-68-2
  • Appearance: White to yellow solid
  • Purity: 98.0%
  • For research use only
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-41790 Supplier Medchemexpress LLC Supplier No. 250MGHY128745
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucosamic acid is an endogenous metabolite with a molecular weight of 195.17 and a chemical formula of C6H13NO6. It is a solid, typically appearing white to yellow, and is primarily used for research applications.

  • Endogenous metabolite
  • Molecular formula: C6H13NO6
  • Molecular weight: 195.17
  • CAS number: 3646-68-2
  • Appearance: White to yellow solid
  • Purity: 98.0%
  • For research use only
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.