Learn More
Medchemexpress LLC D-Gluconic acid, 6-(dihydrogen phosphate), sodium salt (1:3) | 53411-70-4 | 98.0% | 342.08 | 25 MG
SDP
Catalog No. 5000453248
Encompass_Preferred
6-Phosphogluconic acid trisodium is a potent and competitive phosphoglucose isomerase (PGI) inhibitor, exhibiting Kis values of 48 μM for glucose 6-phosphate and 42 μM for fructose 6-phosphate. It is also an endogenous metabolite.
  • Completely inhibits the activity of PGI when glucose 6-phosphate is used as the substrate at concentrations ranging from 0.5 to 2.0 mM.
Catalog No. 50-004-53248 Supplier Medchemexpress LLC Supplier No. 25MGHYW011184
Add to Cart
Add to Cart
6-Phosphogluconic acid trisodium is a potent and competitive phosphoglucose isomerase (PGI) inhibitor, exhibiting Kis values of 48 μM for glucose 6-phosphate and 42 μM for fructose 6-phosphate. It is also an endogenous metabolite.
  • Completely inhibits the activity of PGI when glucose 6-phosphate is used as the substrate at concentrations ranging from 0.5 to 2.0 mM.