Learn More
Abcam D-Gluconic acid, methyl ester, 50MG
Catalog No. 502682804
Encompass_Preferred
MW 210.18 g/mol. Membrane permeant analog of D-gluconic acid. Useful as a component of cell culture media; enzymatic degradation of xylan and glucomannan; various pharmaceutical preparations; as well as an intermediate in carbohydrate based synthetic preparations of sugar derivatives.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2804 Supplier Abcam Supplier No. AB27533450MG
MW 210.18 g/mol. Membrane permeant analog of D-gluconic acid. Useful as a component of cell culture media; enzymatic degradation of xylan and glucomannan; various pharmaceutical preparations; as well as an intermediate in carbohydrate based synthetic preparations of sugar derivatives.
The product is subject to the following: Abcam Restricted Use Statement