Learn More
Alkali Scientific D Gluconic Acid Potassium Salt, 1 Kg
SDP
Catalog No. 5000661578
Encompass_Preferred
Gluconic acid potassium salt is commonly used as a buffer component in biochemical research and laboratory applications. This 1kg reagent plays a crucial role in maintaining stable pH levels in solutions, particularly in processes that require precise control over acidity. The potassium salt of gluconic acid is often employed in microbial culture media, particularly for enhancing cell growth in laboratory settings. Its effectiveness as a buffer stems from its ability to stabilize pH in environments where other buffers might be too acidic or too alkaline. This compound is frequently used in various chemical assays, fermentation studies, and in the production of biochemical reagents.
Catalog No. 50-006-61578 Supplier Alkali Scientific Supplier No. DGL001CP
Gluconic acid potassium salt is commonly used as a buffer component in biochemical research and laboratory applications. This 1kg reagent plays a crucial role in maintaining stable pH levels in solutions, particularly in processes that require precise control over acidity. The potassium salt of gluconic acid is often employed in microbial culture media, particularly for enhancing cell growth in laboratory settings. Its effectiveness as a buffer stems from its ability to stabilize pH in environments where other buffers might be too acidic or too alkaline. This compound is frequently used in various chemical assays, fermentation studies, and in the production of biochemical reagents.