Learn More
Medchemexpress LLC D-Gluconic acid sodium salt | 527-07-1 | MFCD00064210 | ≥98.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB1092A10G
Encompass_Preferred
Gluconate sodium is an orally active glucose derivative that reduces nitric oxide and inflammatory cytokines, inhibits ERK phosphorylation, and possesses antioxidant and antiplatelet activation activities. It demonstrates antitumor activity against colorectal cancer and can improve osteoarthritis, intestinal damage, and acute lung injury.
  • Reduces nitric oxide and inflammatory cytokines
  • Inhibits ERK phosphorylation
  • Possesses antioxidant and antiplatelet activation activities
  • Exhibits antitumor activity against colorectal cancer
  • Improves osteoarthritis symptoms and cartilage damage
  • Alleviates intestinal injury
  • Protects against acute lung injury
Catalog No. 50-004-14060