Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich D-Gluconic Acid Solution

Catalog No. G19514KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G19514KG 25 g
G195125G 1 kg
G19511KG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G19514KG Supplier Sigma Aldrich Supplier No. G19514KG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

D-Gluconic acid is a non-volatile, organic acid generally used as a sequestering agent and as a scale inhibitor. Due to its tendency to chelate with earth alkaline elements and heavy metals, it is also used as a metal extractant. Gluconic acid and its salts areused in hygienic products, food and pharmaceutical industry.

Specifications

CAS 526-95-4
Density 1.234 g/mL at 25°C
Molecular Formula C6H12O7
Refractive Index n20/D 1.4161
Linear Formula C6H12O7
MDL Number MFCD00004240
Quantity 25 g
Synonym 2,3,4,5,6-Pentahydroxycaproic acid
Molecular Weight (g/mol) 196.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.