Learn More
Medchemexpress LLC D-gluconimidic acid, 2-(acetylamino)-2-deoxy-N-phenylcarbamate (Z) | 132489-69-1 | MFCD00145022 | 99.1% | 353.33 | C15H19N3O7 | 10 MG
SDP
Catalog No. 5000242619
Encompass_Preferred
(Z)-PUGNAc is a potent O-GlcNAcase inhibitor used in biochemical and cellular research to modulate protein O-GlcNAcylation. It is supplied as a solid for laboratory research use only and is not intended for human or clinical applications.
  • High purity (99.1%) for reliable experimental results.
  • Molecular weight 353.33 g/mol and formula C15H19N3O7.
  • Off-white to light yellow solid appearance.
  • Store powder at -20°C; store solutions at -80°C, short-term storage at -20°C.
  • Available in 10 MG packaging for small-scale studies.
Catalog No. 50-002-42619 Supplier Medchemexpress LLC Supplier No. HY10824110MG
(Z)-PUGNAc is a potent O-GlcNAcase inhibitor used in biochemical and cellular research to modulate protein O-GlcNAcylation. It is supplied as a solid for laboratory research use only and is not intended for human or clinical applications.
  • High purity (99.1%) for reliable experimental results.
  • Molecular weight 353.33 g/mol and formula C15H19N3O7.
  • Off-white to light yellow solid appearance.
  • Store powder at -20°C; store solutions at -80°C, short-term storage at -20°C.
  • Available in 10 MG packaging for small-scale studies.