Learn More
Medchemexpress LLC β-D-Glucopyranose, 1,2,3,4,6-pentaacetate | 604-69-3 | 98.0% | 390.34 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHY22306
Encompass_Preferred
β-D-Glucose Pentaacetate (Penta-O-acetyl-β-D-glucopyranose) is a glucose derivative primarily used to investigate the mechanism of insulin secretion. It is intended for research use only and not for sale to patients.
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • Structure classification: Saccharides
Catalog No. 50-004-02401