Learn More
Medchemexpress LLC β-D-glucopyranose 1-phosphate (disodium salt) | 83833-15-2 | 99.3% | 304.10 g·mol⁻¹ | C6H11Na2O9P | 5 MG
SDP
Catalog No. 5000283223
Encompass_Preferred
β-D-Glucopyranose 1-phosphate disodium salt is a biochemical reagent used in life-science research as a phosphorylated glucose intermediate. It is supplied as a white to off-white powder in small quantities for analytical and enzymology applications, and is accompanied by batch-specific quality documentation.
  • High chemical purity suitable for analytical use.
  • Molecular formula C6H11Na2O9P; molecular weight 304.10 g·mol⁻¹.
  • Available in small pack sizes: 1 mg, 5 mg, 10 mg.
  • Store sealed at -20°C, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
  • Batch-specific certificate of analysis provided for purity verification.
Catalog No. 50-002-83223 Supplier Medchemexpress LLC Supplier No. HYW1456265MG
β-D-Glucopyranose 1-phosphate disodium salt is a biochemical reagent used in life-science research as a phosphorylated glucose intermediate. It is supplied as a white to off-white powder in small quantities for analytical and enzymology applications, and is accompanied by batch-specific quality documentation.
  • High chemical purity suitable for analytical use.
  • Molecular formula C6H11Na2O9P; molecular weight 304.10 g·mol⁻¹.
  • Available in small pack sizes: 1 mg, 5 mg, 10 mg.
  • Store sealed at -20°C, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
  • Batch-specific certificate of analysis provided for purity verification.