Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-Glucopyranose, 1-thio-, 1-[N-(sulfooxy)-1H-indole-3-ethanimidate], potassium salt (1:1) | 143231-38-3 | 1 MG
SDP

Catalog No. 5000419630
Encompass_Preferred

Glucobrassicin potassium is a glucosinolate (GSL) isolated from wild carrot (Raphanus raphanistrum L.). It can be used in biological fumigation or allelopathy research for weed management.

  • Isolated from wild carrot (Raphanus raphanistrum L.)
  • Suitable for biological fumigation research
  • Suitable for allelopathy research for weed management
  • Glucosinolate (GSL)
  • Purity: 95.97%
  • Appearance: Pale purple to light pink solid
  • Soluble in H2O: 125 mg/mL
  • For research use only

Catalog No. 50-004-19630 Supplier Medchemexpress LLC Supplier No. HYN7659A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glucobrassicin potassium is a glucosinolate (GSL) isolated from wild carrot (Raphanus raphanistrum L.). It can be used in biological fumigation or allelopathy research for weed management.

  • Isolated from wild carrot (Raphanus raphanistrum L.)
  • Suitable for biological fumigation research
  • Suitable for allelopathy research for weed management
  • Glucosinolate (GSL)
  • Purity: 95.97%
  • Appearance: Pale purple to light pink solid
  • Soluble in H2O: 125 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.