Learn More
Medchemexpress LLC β-D-glucopyranose, 1-thio-, 2,3,4,6-tetraacetate | 19879-84-6 | MFCD00063271 | 99.9% | 364.37 g/mol | C14H20O9S | 100 MG
SDP
Catalog No. 5000301959
Encompass_Preferred
1-Thio-beta-D-glucose tetraacetate is the tetraacetylated thio-derivative of beta-D-glucose used as a biochemical reagent and synthetic intermediate in carbohydrate chemistry. It is supplied with high reported purity and supporting analytical documentation, making it suitable for research, method development, and bench-scale syntheses.
  • High purity suitable for analytical and synthetic use.
  • Documented certificate of analysis and safety data sheet available.
  • Stable, isolable solid convenient for handling and storage.
  • Serves as a thioglycosyl donor for glycosylation reactions.
  • Compatible with common organic solvents used in synthesis workflows.
Catalog No. 50-003-01959 Supplier Medchemexpress LLC Supplier No. HYW039865100MG
1-Thio-beta-D-glucose tetraacetate is the tetraacetylated thio-derivative of beta-D-glucose used as a biochemical reagent and synthetic intermediate in carbohydrate chemistry. It is supplied with high reported purity and supporting analytical documentation, making it suitable for research, method development, and bench-scale syntheses.
  • High purity suitable for analytical and synthetic use.
  • Documented certificate of analysis and safety data sheet available.
  • Stable, isolable solid convenient for handling and storage.
  • Serves as a thioglycosyl donor for glycosylation reactions.
  • Compatible with common organic solvents used in synthesis workflows.