Learn More
Medchemexpress LLC β-D-Glucopyranose, 2-(acetylamino)-2-deoxy-, 1,3,4,6-tetraacetate | 7772-79-4 | ≥97.0% | 389.35 | 25 G
SDP
Catalog No. 5000366634
Encompass_Preferred
β-D-Glucopyranose, 2-(acetylamino)-2-deoxy-, 1,3,4,6-tetraacetate is a biochemical reagent used in glycobiology research. Glycobiology involves the study of the structure, synthesis, biology, and evolution of sugars, including carbohydrate chemistry, enzymology of glycan formation and degradation, protein-glycan recognition, and the role of glycans in biological systems. This field is related to basic research, biomedicine, and biotechnology.
  • Biochemical reagent for glycobiology research.
  • Supports studies in carbohydrate chemistry and glycan enzymology.
  • Useful for protein-glycan recognition research.
  • Related to basic research, biomedicine, and biotechnology.
  • Exhibits antiproliferative activity against DBA/2J mouse friend erythroleukemia cells with an IC50 of 400 μM.
Catalog No. 50-003-66634 Supplier Medchemexpress LLC Supplier No. 25GHYW009020
β-D-Glucopyranose, 2-(acetylamino)-2-deoxy-, 1,3,4,6-tetraacetate is a biochemical reagent used in glycobiology research. Glycobiology involves the study of the structure, synthesis, biology, and evolution of sugars, including carbohydrate chemistry, enzymology of glycan formation and degradation, protein-glycan recognition, and the role of glycans in biological systems. This field is related to basic research, biomedicine, and biotechnology.
  • Biochemical reagent for glycobiology research.
  • Supports studies in carbohydrate chemistry and glycan enzymology.
  • Useful for protein-glycan recognition research.
  • Related to basic research, biomedicine, and biotechnology.
  • Exhibits antiproliferative activity against DBA/2J mouse friend erythroleukemia cells with an IC50 of 400 μM.