Learn More
Medchemexpress LLC β-D-Glucopyranose, 4-O-(2,3,4,6-tetra-O-acetyl-β-D-galactopyranosyl)-, 1,2,3,6-tetraacetate | 6291-42-5 | 98.0% | 678.59 | 10 G
SDP
Catalog No. 5000451304
Encompass_Preferred
Lactose octaacetate is a white to off-white solid powder that exhibits mild to moderate antifungal activity against some fungi, while showing low or no activity against bacteria and yeast. It also demonstrates low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and possesses antiviral activity against PV-1.
  • Mild to moderate antifungal activity
  • Low or no activity against bacteria and yeast
  • Low cytotoxicity to MDBK cells, Hep-2, and MDCK cells
  • Antiviral activity against PV-1
Catalog No. 50-004-51304 Supplier Medchemexpress LLC Supplier No. HYW00886410G
Lactose octaacetate is a white to off-white solid powder that exhibits mild to moderate antifungal activity against some fungi, while showing low or no activity against bacteria and yeast. It also demonstrates low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and possesses antiviral activity against PV-1.
  • Mild to moderate antifungal activity
  • Low or no activity against bacteria and yeast
  • Low cytotoxicity to MDBK cells, Hep-2, and MDCK cells
  • Antiviral activity against PV-1