Learn More
Medchemexpress LLC β-D-Glucopyranose, 4-O-(2,3,4,6-tetra-O-acetyl-β-D-galactopyranosyl)-, 1,2,3,6-tetraacetate | 6291-42-5 | 98.0% | 678.59 | 5 G
SDP
Catalog No. 5000453741
Encompass_Preferred
Lactose octaacetate demonstrates mild to moderate antifungal activity against certain fungi, but it exhibits low or no activity against bacteria and yeast. It also shows low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and has antiviral activity against PV-1.
  • Mild to moderate antifungal activity against some fungi.
  • Low or no activity against bacteria and yeast.
  • Low cytotoxicity to MDBK cells, Hep-2, and MDCK cells.
  • Antiviral activity against PV-1.
Catalog No. 50-004-53741 Supplier Medchemexpress LLC Supplier No. HYW0088645G
Lactose octaacetate demonstrates mild to moderate antifungal activity against certain fungi, but it exhibits low or no activity against bacteria and yeast. It also shows low cytotoxicity to MDBK cells, Hep-2, and MDCK cells, and has antiviral activity against PV-1.
  • Mild to moderate antifungal activity against some fungi.
  • Low or no activity against bacteria and yeast.
  • Low cytotoxicity to MDBK cells, Hep-2, and MDCK cells.
  • Antiviral activity against PV-1.