Learn More
Medchemexpress LLC β-D-Glucopyranoside, 2-(3,4-dihydroxyphenyl)ethyl, 6-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoate] | 105471-98-5 | MFCD22125003 | 99.9% | 478.45 | C23H26O11 | 10 MG
SDP
Catalog No. 5000295724
Encompass_Preferred
Calceolarioside B is a naturally occurring phenylethanoid glycoside supplied as a research-grade small-molecule. It is reported to inhibit RLAR activity and has shown antiviral, immunomodulatory, and anticancer activity in in vitro assays. Use is limited to laboratory research only.
  • Phenylethanoid glycoside chemical class with defined structure and formula.
  • Reported RLAR inhibitor (IC50 ≈ 23.99 μM) useful for enzyme assays.
  • Shown to reduce IL-6 levels and inhibit viral entry in vitro.
  • High reported purity suitable for biochemical and cell-based studies.
  • Supplied in small research pack sizes for laboratory use.
Catalog No. 50-002-95724 Supplier Medchemexpress LLC Supplier No. HYN053910MG
Calceolarioside B is a naturally occurring phenylethanoid glycoside supplied as a research-grade small-molecule. It is reported to inhibit RLAR activity and has shown antiviral, immunomodulatory, and anticancer activity in in vitro assays. Use is limited to laboratory research only.
  • Phenylethanoid glycoside chemical class with defined structure and formula.
  • Reported RLAR inhibitor (IC50 ≈ 23.99 μM) useful for enzyme assays.
  • Shown to reduce IL-6 levels and inhibit viral entry in vitro.
  • High reported purity suitable for biochemical and cell-based studies.
  • Supplied in small research pack sizes for laboratory use.