Learn More
Medchemexpress LLC β-D-glucopyranoside, dodecyl | 59122-55-3 | MFCD00063298 | 98.0% | 348.47 | C18H36O6 | 1 G
SDP
Catalog No. 5000282919
Encompass_Preferred
Dodecyl β-D-glucopyranoside is a non-ionic detergent and surfactant commonly used to solubilize and purify membrane proteins and for other biochemical research applications. It forms micelles above its critical micelle concentration and can interact with proteins, affecting fluorescence measurements; it is supplied as a high-purity laboratory reagent.
Catalog No. 50-002-82919 Supplier Medchemexpress LLC Supplier No. HYW1426921G
Dodecyl β-D-glucopyranoside is a non-ionic detergent and surfactant commonly used to solubilize and purify membrane proteins and for other biochemical research applications. It forms micelles above its critical micelle concentration and can interact with proteins, affecting fluorescence measurements; it is supplied as a high-purity laboratory reagent.