Learn More
Medchemexpress LLC α-D-glucopyranoside, ethyl 1-thio, 2,3,4,6-tetraacetate | 41670-79-5 | MFCD00797608 | >97.0% | 392.42 g/mol | C16H24O9S | 10 G
SDP
Catalog No. 5000299451
Encompass_Preferred
α-D-Glucopyranoside, ethyl 1-thio,2,3,4,6-tetraacetate is a tetraacetylated ethyl 1-thio derivative of D-glucose (CAS 41670-79-5) used as a protected thio-glycoside intermediate in carbohydrate chemistry. It is supplied at high purity for use in glycosylation reactions, synthetic building-block preparation, and analytical studies.
  • Protected thio-glycoside suitable for glycosylation and carbohydrate synthesis.
  • Tetraacetylation improves stability and compatibility with organic solvents.
  • High purity (>97%) for consistent synthetic results.
  • Molecular formula C16H24O9S and molecular weight 392.42 g/mol.
  • Available in laboratory pack sizes for synthetic workflows.
Catalog No. 50-002-99451 Supplier Medchemexpress LLC Supplier No. HYW03984810G
α-D-Glucopyranoside, ethyl 1-thio,2,3,4,6-tetraacetate is a tetraacetylated ethyl 1-thio derivative of D-glucose (CAS 41670-79-5) used as a protected thio-glycoside intermediate in carbohydrate chemistry. It is supplied at high purity for use in glycosylation reactions, synthetic building-block preparation, and analytical studies.
  • Protected thio-glycoside suitable for glycosylation and carbohydrate synthesis.
  • Tetraacetylation improves stability and compatibility with organic solvents.
  • High purity (>97%) for consistent synthetic results.
  • Molecular formula C16H24O9S and molecular weight 392.42 g/mol.
  • Available in laboratory pack sizes for synthetic workflows.