Learn More
Medchemexpress LLC α-D-glucopyranoside, ethyl 1-thio-2,3,4,6-tetraacetate | 41670-79-5 | MFCD00797608 | >97.0% | 392.42 g/mol | C16H24O9S | 500 MG
SDP
Catalog No. 5000301960
Encompass_Preferred
α-D-Glucopyranoside, ethyl 1-thio-2,3,4,6-tetraacetate is an acetylated thioglycoside reagent used in carbohydrate chemistry and glycobiology research. It serves as a synthetic intermediate and assay substrate supplied in laboratory-scale quantities for analytical and preparative workflows.
  • Acetylated thioglycoside useful as a synthetic intermediate in carbohydrate chemistry.
  • Suitable for glycobiology assays and analytical method development.
  • Supplied in small laboratory quantities for research use.
  • Requires storage and handling per SDS and COA to maintain integrity.
  • Identified by CAS 41670-79-5 for unambiguous compound reference.
Catalog No. 50-003-01960 Supplier Medchemexpress LLC Supplier No. HYW039848500MG
α-D-Glucopyranoside, ethyl 1-thio-2,3,4,6-tetraacetate is an acetylated thioglycoside reagent used in carbohydrate chemistry and glycobiology research. It serves as a synthetic intermediate and assay substrate supplied in laboratory-scale quantities for analytical and preparative workflows.
  • Acetylated thioglycoside useful as a synthetic intermediate in carbohydrate chemistry.
  • Suitable for glycobiology assays and analytical method development.
  • Supplied in small laboratory quantities for research use.
  • Requires storage and handling per SDS and COA to maintain integrity.
  • Identified by CAS 41670-79-5 for unambiguous compound reference.