Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-glucopyranosiduronic acid, 4-formyl-2-nitrophenyl, methyl ester, 2,3,4-triacetate | 148579-93-5 | 98.6% | 483.38 g/mol | C20H21NO13 | 250 MG
SDP

Catalog No. 5000202378
Encompass_Preferred

Me-triacetyl-β-D-glucopyranuronate-Ph-ald-NO2 is a cleavable antibody-drug conjugate (ADC) linker used in the synthesis of ADCs. The material is provided as a triacetylated methyl ester derivative with reported high purity, DMSO solubility, and specified storage recommendations.

  • Cleavable glucuronide-based ADC linker.
  • High purity (98.6% by LCMS).
  • Soluble in DMSO 200 mg/mL; may require ultrasonic and use of newly opened DMSO.
  • Molecular weight 483.38 g/mol.
  • Chemical formula C20H21NO13.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-02378 Supplier Medchemexpress LLC Supplier No. HY131086250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Me-triacetyl-β-D-glucopyranuronate-Ph-ald-NO2 is a cleavable antibody-drug conjugate (ADC) linker used in the synthesis of ADCs. The material is provided as a triacetylated methyl ester derivative with reported high purity, DMSO solubility, and specified storage recommendations.

  • Cleavable glucuronide-based ADC linker.
  • High purity (98.6% by LCMS).
  • Soluble in DMSO 200 mg/mL; may require ultrasonic and use of newly opened DMSO.
  • Molecular weight 483.38 g/mol.
  • Chemical formula C20H21NO13.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.