Learn More
Medchemexpress LLC β-D-glucopyranosiduronic acid derivative (p-nitrophenyl carbonate methyl ester) | 894095-98-8 | MFCD31715405 | 99.2% | 913.83 | C45H43N3O18 | 100 MG
SDP
Catalog No. 5000204235
Encompass_Preferred
β-D-glucuronide-pNP-carbonate is a cleavable antibody-drug conjugate (ADC) linker used as a building block in the synthesis of ADCs. It features a p-nitrophenyl (pNP) carbonate leaving group attached to a β-D-glucuronide moiety, enabling enzymatic or chemical cleavage to release payloads for targeted delivery. Intended for research use only.
  • Cleavable glycosidase-sensitive linker for controlled payload release.
  • p-Nitrophenyl carbonate activation enables facile coupling to amines.
  • High purity suitable for synthetic and medicinal chemistry workflows.
  • Available as a solid reagent in small research quantities for laboratory use.
Catalog No. 50-002-04235 Supplier Medchemexpress LLC Supplier No. HY136329100MG
β-D-glucuronide-pNP-carbonate is a cleavable antibody-drug conjugate (ADC) linker used as a building block in the synthesis of ADCs. It features a p-nitrophenyl (pNP) carbonate leaving group attached to a β-D-glucuronide moiety, enabling enzymatic or chemical cleavage to release payloads for targeted delivery. Intended for research use only.
  • Cleavable glycosidase-sensitive linker for controlled payload release.
  • p-Nitrophenyl carbonate activation enables facile coupling to amines.
  • High purity suitable for synthetic and medicinal chemistry workflows.
  • Available as a solid reagent in small research quantities for laboratory use.