Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-glucopyranosiduronic acid methyl ester, 2,3,4-triacetate | 34213-34-8 | MFCD09841789 | 348.30 g/mol | C14H20O10 | 10 G
SDP

Catalog No. 5000299825
Encompass_Preferred

β-D-Glucopyranosiduronic acid, methyl ester, 2,3,4-triacetate is the triacetylated methyl ester derivative of β-D-glucuronic acid used as a biochemical assay reagent and reference standard in carbohydrate chemistry and analytical method development.

  • Used as a biochemical assay reagent and reference standard.
  • Triacetylated methyl ester derivative of β-D-glucuronic acid.
  • Molecular formula C14H20O10; molecular weight 348.30 g/mol.
  • Powder stability: store at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Stability in solvent: store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-99825 Supplier Medchemexpress LLC Supplier No. HYW03988710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Glucopyranosiduronic acid, methyl ester, 2,3,4-triacetate is the triacetylated methyl ester derivative of β-D-glucuronic acid used as a biochemical assay reagent and reference standard in carbohydrate chemistry and analytical method development.

  • Used as a biochemical assay reagent and reference standard.
  • Triacetylated methyl ester derivative of β-D-glucuronic acid.
  • Molecular formula C14H20O10; molecular weight 348.30 g/mol.
  • Powder stability: store at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Stability in solvent: store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.