Learn More
Medchemexpress LLC β-D-glucopyranosiduronic acid p-nitrophenyl carbonate derivative | 894095-98-8 | 99.6% | 913.83 g/mol | C45H43N3O18 | 1 G
SDP
Catalog No. 5000203790
Encompass_Preferred
β-D-glucuronide-pNP-carbonate is a glucuronide-cleavable carbonate linker used to attach payloads to antibodies for research-scale antibody-drug conjugate (ADC) synthesis. It is supplied as a purified small-molecule reagent for linker-payload assembly and analytical characterization.
  • Cleavable glucuronide-based linker responsive to glycosidase cleavage.
  • Reported purity around 99.6%.
  • Molecular formula C45H43N3O18; molecular weight 913.83 g/mol.
  • Available in small-scale packaging including 1 G; larger quantities available on request.
  • Supporting documents include datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-03790 Supplier Medchemexpress LLC Supplier No. HY1363291G
β-D-glucuronide-pNP-carbonate is a glucuronide-cleavable carbonate linker used to attach payloads to antibodies for research-scale antibody-drug conjugate (ADC) synthesis. It is supplied as a purified small-molecule reagent for linker-payload assembly and analytical characterization.
  • Cleavable glucuronide-based linker responsive to glycosidase cleavage.
  • Reported purity around 99.6%.
  • Molecular formula C45H43N3O18; molecular weight 913.83 g/mol.
  • Available in small-scale packaging including 1 G; larger quantities available on request.
  • Supporting documents include datasheet, certificate of analysis, and safety data sheet.