Learn More
Medchemexpress LLC β-D-Glucopyranosyl abscisate | 21414-42-6 | 95.0% | 426.46 | 1 MG
SDP
Catalog No. 5000438234
Encompass_Preferred
β-D-Glucopyranosyl abscisate (ABA-GE) is a hydrolyzable abscisic acid (ABA) conjugate that accumulates in the vacuole and presumably also in the endoplasmic reticulum. The deconjugation of β-D-Glucopyranosyl abscisate allows the rapid formation of free ABA in response to abiotic stress conditions such as dehydration and salt stress. β-D-Glucopyranosyl abscisate contributes to the maintenance of ABA homeostasis.
  • Hydrolyzable abscisic acid (ABA) conjugate.
  • Accumulates in the vacuole and endoplasmic reticulum.
  • Deconjugation leads to rapid formation of free ABA under abiotic stress (dehydration, salt stress).
  • Contributes to the maintenance of ABA homeostasis.
Catalog No. 50-004-38234 Supplier Medchemexpress LLC Supplier No. HY1119741MG
β-D-Glucopyranosyl abscisate (ABA-GE) is a hydrolyzable abscisic acid (ABA) conjugate that accumulates in the vacuole and presumably also in the endoplasmic reticulum. The deconjugation of β-D-Glucopyranosyl abscisate allows the rapid formation of free ABA in response to abiotic stress conditions such as dehydration and salt stress. β-D-Glucopyranosyl abscisate contributes to the maintenance of ABA homeostasis.
  • Hydrolyzable abscisic acid (ABA) conjugate.
  • Accumulates in the vacuole and endoplasmic reticulum.
  • Deconjugation leads to rapid formation of free ABA under abiotic stress (dehydration, salt stress).
  • Contributes to the maintenance of ABA homeostasis.