Learn More
Medchemexpress LLC β-D-Glucopyranosyl abscisate | 21414-42-6 | 95.0% | 426.46 | 50 MG
SDP
Catalog No. 5000438994
Encompass_Preferred
β-D-Glucopyranosyl abscisate (ABA-GE) is a hydrolyzable abscisic acid (ABA) conjugate that accumulates in the vacuole and endoplasmic reticulum. It allows for the rapid formation of free ABA in response to abiotic stress conditions like dehydration and salt stress, contributing to the maintenance of ABA homeostasis.
  • Facilitates rapid free ABA formation during abiotic stress
  • Maintains ABA homeostasis as a major ABA catabolite exported from the cytosol
  • Vacuolar transport mediated by ATP-binding cassette and proton-antiport mechanisms in arabidopsis
Catalog No. 50-004-38994 Supplier Medchemexpress LLC Supplier No. HY11197450MG
β-D-Glucopyranosyl abscisate (ABA-GE) is a hydrolyzable abscisic acid (ABA) conjugate that accumulates in the vacuole and endoplasmic reticulum. It allows for the rapid formation of free ABA in response to abiotic stress conditions like dehydration and salt stress, contributing to the maintenance of ABA homeostasis.
  • Facilitates rapid free ABA formation during abiotic stress
  • Maintains ABA homeostasis as a major ABA catabolite exported from the cytosol
  • Vacuolar transport mediated by ATP-binding cassette and proton-antiport mechanisms in arabidopsis