Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-glucopyranosyl abscisate | 21414-42-6 | MFCD30748334 | 95.0% | 426.46 g/mol | C21H30O9 | 5 MG
SDP

Catalog No. 5000264251
Encompass_Preferred

β-D-Glucopyranosyl abscisate is the beta-D-glucosyl ester of abscisic acid, used as a research reagent in plant biology and biochemical studies. Supplied as a characterized solid, it is intended for laboratory research applications where abscisic acid conjugates are studied.

  • Purity 95.04% (manufacturer COA).
  • Physical form solid, white to off-white.
  • Molecular weight 426.46 g/mol.
  • Molecular formula C21H30O9.
  • Suitable for plant physiology and biochemical assays.
  • Storage protect from light and store under nitrogen at -20°C; in solvent: -80°C up to 6 months.

Catalog No. 50-002-64251 Supplier Medchemexpress LLC Supplier No. HY1119745MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Glucopyranosyl abscisate is the beta-D-glucosyl ester of abscisic acid, used as a research reagent in plant biology and biochemical studies. Supplied as a characterized solid, it is intended for laboratory research applications where abscisic acid conjugates are studied.

  • Purity 95.04% (manufacturer COA).
  • Physical form solid, white to off-white.
  • Molecular weight 426.46 g/mol.
  • Molecular formula C21H30O9.
  • Suitable for plant physiology and biochemical assays.
  • Storage protect from light and store under nitrogen at -20°C; in solvent: -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.