Learn More
Medchemexpress LLC β-D-Glucopyranosyl abscisate (ABA-GE) | 21414-42-6 | 95.04% | 426.46 | 100 MG
SDP
Catalog No. 5000439352
Encompass_Preferred
β-D-Glucopyranosyl abscisate (ABA-GE) is a hydrolyzable abscisic acid (ABA) conjugate that accumulates in the vacuole and presumably also in the endoplasmic reticulum. The deconjugation of β-D-Glucopyranosyl abscisate allows the rapid formation of free ABA in response to abiotic stress conditions such as dehydration and salt stress. β-D-Glucopyranosyl abscisate contributes to the maintenance of ABA homeostasis. For research use only.
  • Accumulates in the vacuole and endoplasmic reticulum.
  • Allows rapid formation of free ABA in response to abiotic stress conditions like dehydration and salt stress.
  • Contributes to the maintenance of ABA homeostasis.
  • Vacuolar transport is mediated by ATP-binding cassette and proton-antiport mechanisms.
Catalog No. 50-004-39352 Supplier Medchemexpress LLC Supplier No. HY111974100MG
β-D-Glucopyranosyl abscisate (ABA-GE) is a hydrolyzable abscisic acid (ABA) conjugate that accumulates in the vacuole and presumably also in the endoplasmic reticulum. The deconjugation of β-D-Glucopyranosyl abscisate allows the rapid formation of free ABA in response to abiotic stress conditions such as dehydration and salt stress. β-D-Glucopyranosyl abscisate contributes to the maintenance of ABA homeostasis. For research use only.
  • Accumulates in the vacuole and endoplasmic reticulum.
  • Allows rapid formation of free ABA in response to abiotic stress conditions like dehydration and salt stress.
  • Contributes to the maintenance of ABA homeostasis.
  • Vacuolar transport is mediated by ATP-binding cassette and proton-antiport mechanisms.