Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-D-glucopyranuronic acid, methyl ester, 1,2,3,4-tetrakis(2-methylpropanoate) | 150607-94-6 | 98.1% | 488.53 g·mol⁻¹ | C23H36O11 | 25 MG
SDP

Catalog No. 5000281490
Encompass_Preferred

β-D-Glucopyranuronic acid, methyl ester, 1,2,3,4-tetrakis(2-methylpropanoate) is a research-grade glucuronic acid derivative supplied as a small-quantity biochemical reagent for glycobiology and carbohydrate chemistry. It is intended for use in structural studies, synthetic chemistry, and enzymology assays where protected glucuronic acid derivatives are required.

  • Purity 98.1%.
  • Molecular weight 488.53 g·mol⁻¹.
  • Chemical formula C23H36O11.
  • Available in a 25 mg pack size.
  • Suitable for glycobiology and carbohydrate chemistry research.
  • Soluble in dimethyl sulfoxide (DMSO) for preparation of stock solutions.

Catalog No. 50-002-81490 Supplier Medchemexpress LLC Supplier No. HYW40228625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Glucopyranuronic acid, methyl ester, 1,2,3,4-tetrakis(2-methylpropanoate) is a research-grade glucuronic acid derivative supplied as a small-quantity biochemical reagent for glycobiology and carbohydrate chemistry. It is intended for use in structural studies, synthetic chemistry, and enzymology assays where protected glucuronic acid derivatives are required.

  • Purity 98.1%.
  • Molecular weight 488.53 g·mol⁻¹.
  • Chemical formula C23H36O11.
  • Available in a 25 mg pack size.
  • Suitable for glycobiology and carbohydrate chemistry research.
  • Soluble in dimethyl sulfoxide (DMSO) for preparation of stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.