Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-d-glucopyranuronic acid, methyl ester, 1,2,3,4-tetrakis(2-methylpropanoate) | 150607-94-6 | 98.1% | 488.53 g·mol⁻¹ | C23H36O11 | 5 MG
SDP

Catalog No. 5000281507
Encompass_Preferred

β-D-Glucopyranuronic acid methyl ester, 1,2,3,4-tetrakis(2-methylpropanoate) is a synthetic carbohydrate derivative supplied as a high-purity biochemical reagent for laboratory research. It is intended for glycobiology and carbohydrate chemistry applications and is provided in small milligram quantities for assay development and analytical studies. Not for human or animal diagnostic or therapeutic use.

  • High purity: 98.1%.
  • Molecular formula: C23H36O11.
  • Molecular weight: 488.53 g·mol⁻¹.
  • CAS number: 150607-94-6.
  • Available in small milligram packs such as 5 MG.
  • Suitable for biochemical assay and glycobiology research.
  • Intended for research use only; not for clinical or food use.

Catalog No. 50-002-81507 Supplier Medchemexpress LLC Supplier No. HYW4022865MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Glucopyranuronic acid methyl ester, 1,2,3,4-tetrakis(2-methylpropanoate) is a synthetic carbohydrate derivative supplied as a high-purity biochemical reagent for laboratory research. It is intended for glycobiology and carbohydrate chemistry applications and is provided in small milligram quantities for assay development and analytical studies. Not for human or animal diagnostic or therapeutic use.

  • High purity: 98.1%.
  • Molecular formula: C23H36O11.
  • Molecular weight: 488.53 g·mol⁻¹.
  • CAS number: 150607-94-6.
  • Available in small milligram packs such as 5 MG.
  • Suitable for biochemical assay and glycobiology research.
  • Intended for research use only; not for clinical or food use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.