Learn More
Cayman Chemical D-GlucosamIn-6-sulfate 10mg
SDP
Catalog No. 502993950
Encompass_Preferred
A glycosaminoglycan; activates the glmS ribozyme from B. subtilis (200 µM); has been used to form polyvalent dendrimer conjugates that inhibit angiogenesis, cell proliferation, and scar tissue formation
Catalog No. 50-299-3950 Supplier Cayman Chemical Supplier No. 2309710 MG
A glycosaminoglycan; activates the glmS ribozyme from B. subtilis (200 µM); has been used to form polyvalent dendrimer conjugates that inhibit angiogenesis, cell proliferation, and scar tissue formation