Learn More
Cayman Chemical D-GlucosamIn-6-sulfate 50mg
SDP
Catalog No. 502995077
Encompass_Preferred
A glycosaminoglycan; activates the glmS ribozyme from B. subtilis (200 µM); has been used to form polyvalent dendrimer conjugates that inhibit angiogenesis, cell proliferation, and scar tissue formation
Catalog No. 50-299-5077 Supplier Cayman Chemical Supplier No. 2309750 MG
A glycosaminoglycan; activates the glmS ribozyme from B. subtilis (200 µM); has been used to form polyvalent dendrimer conjugates that inhibit angiogenesis, cell proliferation, and scar tissue formation