Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucosamine 6-phosphate | 3616-42-0 | 259.15 | 25 MG
SDP

Catalog No. 5000389153
Encompass_Preferred

D-Glucosamine 6-phosphate is an endogenous metabolite that can be synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT). It can be used for the research of diabetes mellitus.

  • Purity: 99.97%
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite, human gut microbiota metabolites
  • Solubility (in vitro): H2O: 125 mg/mL (482.35 mM; need ultrasonic)

Catalog No. 50-003-89153 Supplier Medchemexpress LLC Supplier No. 25MGHYN11848
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucosamine 6-phosphate is an endogenous metabolite that can be synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT). It can be used for the research of diabetes mellitus.

  • Purity: 99.97%
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite, human gut microbiota metabolites
  • Solubility (in vitro): H2O: 125 mg/mL (482.35 mM; need ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.