Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucosamine 6-phosphate | 3616-42-0 | 259.15 | 50 MG
SDP

Catalog No. 5000416599
Encompass_Preferred

D-Glucosamine 6-phosphate is an endogenous metabolite that can be synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT). It is utilized for research into diabetes mellitus.

  • Endogenous metabolite
  • Synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT)
  • Applicable for research of diabetes mellitus
  • Molecular weight of 259.15
  • Chemical formula is C6H14NO8P
  • CAS number is 3616-42-0
  • Appears as a white to off-white solid
  • High purity of 99.97%
  • Soluble in H2O at 125 mg/mL
  • Store at -20°C, sealed, away from moisture and light

Catalog No. 50-004-16599 Supplier Medchemexpress LLC Supplier No. HYN11848R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucosamine 6-phosphate is an endogenous metabolite that can be synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT). It is utilized for research into diabetes mellitus.

  • Endogenous metabolite
  • Synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT)
  • Applicable for research of diabetes mellitus
  • Molecular weight of 259.15
  • Chemical formula is C6H14NO8P
  • CAS number is 3616-42-0
  • Appears as a white to off-white solid
  • High purity of 99.97%
  • Soluble in H2O at 125 mg/mL
  • Store at -20°C, sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.