Learn More
Medchemexpress LLC D-glucosamine 6-phosphate | 3616-42-0 | MFCD00067382 | 100.0% | 259.15 g/mol | C6H14NO8P | 10 MG
SDP
Catalog No. 5000295747
Encompass_Preferred
D-Glucosamine 6-phosphate is the 6-phosphorylated form of D-glucosamine and is used as an analytical and biochemical standard for studies of amino-sugar metabolism, glycosaminoglycan biosynthesis, enzymatic assays, and metabolomics. It is provided as a high-purity compound for laboratory research applications.
  • High purity suitable for analytical applications.
  • Useful as a reference standard for LC-MS and HPLC.
  • Compatible with enzymatic and kinetic assays.
  • Supplied in small milligram quantities for laboratory use.
  • Molecular weight 259.15 g/mol; chemical formula C6H14NO8P.
Catalog No. 50-002-95747 Supplier Medchemexpress LLC Supplier No. HYN1184810MG
D-Glucosamine 6-phosphate is the 6-phosphorylated form of D-glucosamine and is used as an analytical and biochemical standard for studies of amino-sugar metabolism, glycosaminoglycan biosynthesis, enzymatic assays, and metabolomics. It is provided as a high-purity compound for laboratory research applications.
  • High purity suitable for analytical applications.
  • Useful as a reference standard for LC-MS and HPLC.
  • Compatible with enzymatic and kinetic assays.
  • Supplied in small milligram quantities for laboratory use.
  • Molecular weight 259.15 g/mol; chemical formula C6H14NO8P.