Learn More
Medchemexpress LLC D-Glucosamine 6-phosphate | 3616-42-0 | MFCD00067382 | 259.15 | 25 MG
SDP
Catalog No. 5000389435
Encompass_Preferred
D-Glucosamine 6-phosphate is an endogenous metabolite that can be synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT). It can be used for the research of diabetes mellitus.
  • Endogenous metabolite
  • For research use only (not for sale to patients)
Catalog No. 50-003-89435 Supplier Medchemexpress LLC Supplier No. 25MGHYN11848R
D-Glucosamine 6-phosphate is an endogenous metabolite that can be synthesized by glutamine fructose-6-phosphate amidotransferase (GFAT). It can be used for the research of diabetes mellitus.
  • Endogenous metabolite
  • For research use only (not for sale to patients)