Learn More
Medchemexpress LLC D-Glucosamine 6-phosphate (Standard) | 3616-42-0 | 259.15 | 1MG
SDP
Catalog No. 5000414628
Encompass_Preferred
D-Glucosamine 6-phosphate (Standard) is a laboratory chemical used in the manufacture of substances. It is stable under recommended storage conditions, which include -20°C in a sealed, dry environment, away from light and moisture. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month. Short-term shipping at room temperature (less than 2 weeks) is acceptable.
  • Classified as a laboratory chemical
  • Used for the manufacture of substances
  • Stable under recommended storage conditions
  • Long-term storage at -20°C in a sealed, dry environment, away from light and moisture
  • Can be stored in solvent at -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature for durations less than 2 weeks
Catalog No. 50-004-14628 Supplier Medchemexpress LLC Supplier No. HYN11848R1MG
D-Glucosamine 6-phosphate (Standard) is a laboratory chemical used in the manufacture of substances. It is stable under recommended storage conditions, which include -20°C in a sealed, dry environment, away from light and moisture. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month. Short-term shipping at room temperature (less than 2 weeks) is acceptable.
  • Classified as a laboratory chemical
  • Used for the manufacture of substances
  • Stable under recommended storage conditions
  • Long-term storage at -20°C in a sealed, dry environment, away from light and moisture
  • Can be stored in solvent at -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature for durations less than 2 weeks