Learn More
Medchemexpress LLC D-(+)-glucosamine hydrochloride | 66-84-2 | 215.63 | 50 MG
SDP
Catalog No. 5000417124
Encompass_Preferred
Glucosamine hydrochloride is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, also used as a dietary supplement. It is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, exerting pharmacological effects on osteoarthritic cartilage and chondrocytes. This product exhibits dose-dependent DPPH antioxidant activity and inhibits HIF-1α at the protein level.
  • Exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Inhibits HIF-1α at the protein level.
  • Decreases phosphorylation of p70S6K and S6, translation-related proteins.
  • Decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.
Catalog No. 50-004-17124 Supplier Medchemexpress LLC Supplier No. HYN0733R50MG
Glucosamine hydrochloride is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids, also used as a dietary supplement. It is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, exerting pharmacological effects on osteoarthritic cartilage and chondrocytes. This product exhibits dose-dependent DPPH antioxidant activity and inhibits HIF-1α at the protein level.
  • Exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Inhibits HIF-1α at the protein level.
  • Decreases phosphorylation of p70S6K and S6, translation-related proteins.
  • Decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.