Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%

Catalog No. AC119900025
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119900025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC119900025 Supplier Thermo Scientific Chemicals Supplier No. 119900025
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H13NO5·HCl
Molecular Weight (g/mol) 215.64

Specifications

Melting Point 190°C to 194°C
Color Brown-Yellow to Yellow
Loss on Drying 1.0 % max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 01,902
Merck Index 10,4318
Specific Rotation Condition + 72.50 (20.00°C c=2,H2O,5hrs.)
Specific Rotation + 72.5
Solubility Information Solubility in water: soluble
Formula Weight 215.64
Percent Purity 98+%
Physical Form Solution
Chemical Name or Material D-Glucosamine hydrochloride
Show More Show Less
EINECSNumber 200-638-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.