Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Glucosamine hydrochloride, 99.83%, MP Biomedicals™

Catalog No. ICN19467125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19467125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19467125 Supplier MP Biomedicals, Inc Supplier No. 0219467125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity cell culture grade chemical for protein N-glycosylation and cell differentiation studies.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H13NO5·HCI
Molecular Weight (g/mol) 215.63
Synonym 2-Amino-2-deoxy-D -glucose, Chitosamine

Specifications

Color White
Assay Percent Range 0.9983
Quantity 25 g
Formula Weight 215.6
Percent Purity ≥99%
Grade Cell Culture
Physical Form Powder
Chemical Name or Material D-(+)-Glucosamine hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.