Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Glucosamine Hydrochloride, 99%, MP Biomedicals™

Catalog No. ICN10178290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10178290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10178290 Supplier MP Biomedicals, Inc Supplier No. 0210178290
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

USP grade D-(+)-glucosamine hydrochloride powder for laboratory research applications.

Molecular Biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H13NO5.ClH
Molecular Weight (g/mol) 215.64
MDL Number MFCD00135831
Synonym 2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride

Specifications

Assay Percent Range 99%
Quantity 500 g
Formula Weight 215.6
Percent Purity 99.8%
Grade Not Available
Physical Form Solid
Chemical Name or Material D-(+)-Glucosamine Hydrochloride
Recommended Storage Store at room temperature (15° to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.