Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Glucosamine Hydrochloride, Spectrum™ Chemical
SDP

Catalog No. 18620803 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
50 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-803 50 kg
18-620-797 25 g
18-620-798 100 g
18-620-799 500 g
18-620-800 1 kg
18-620-801 5 kg
18-620-802 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-620-803 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GL11050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Synonym: Chitosamine Hydrochloride, 2-Amino-2-Deoxy-D-Glucose Hydrochloride, 2-Amino-D-Glucose Hydrochloride

Chitosamine Hydrochloride, 2-Amino-2-Deoxy-D-Glucose Hydrochloride, 2-Amino-D-Glucose Hydrochloride

Chemical Identifiers

CAS 66-84-2

Specifications

Packaging Drum
Quantity 50 kg
Molecular Weight (g/mol) 215.64
Percent Purity 98%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material D-Glucosamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.