Learn More
Medchemexpress LLC D-Glucosamine (sulfate) | 29031-19-4 | 277.25 | 500 MG
SDP
Catalog No. 5000389083
Encompass_Preferred
Glucosamine sulfate (D-Glucosamine sulfate) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids. It is used as a dietary supplement and is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which, when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Used as a dietary supplement.
  • Exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Short-term treatment inhibits HIF-1α at the protein level, decreasing phosphorylation of p70S6K and S6, translation-related proteins.
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.
  • Purity of 98.0%.
Catalog No. 50-003-89083 Supplier Medchemexpress LLC Supplier No. HYN0487500MG
Glucosamine sulfate (D-Glucosamine sulfate) is an amino sugar and a prominent precursor in the biochemical synthesis of glycosylated proteins and lipids. It is used as a dietary supplement and is a natural constituent of glycosaminoglycans in the cartilage matrix and synovial fluid, which, when administered exogenously, exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Used as a dietary supplement.
  • Exerts pharmacological effects on osteoarthritic cartilage and chondrocytes.
  • Exhibits dose-dependent DPPH antioxidant activity.
  • Short-term treatment inhibits HIF-1α at the protein level, decreasing phosphorylation of p70S6K and S6, translation-related proteins.
  • Significantly decreases renal expression of α-smooth muscle actin, collagen I, and fibronectin in obstructed kidneys and TGF-β1-treated renal cells.
  • Purity of 98.0%.