Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-Glucosamine sulfate, 98%

Catalog No. AAJ6627114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6627114 25 g
AAJ6627106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6627114 Supplier Thermo Scientific Chemicals Supplier No. J6627114
Add to Cart

missing translation for 'validMainframeMsg'

Novel application of glucosamine to prepare medical agent for treating vertigo. Found in chitin, in mucoproteins, and in mucopolysaccharides. Antiarthritic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble at 1mg/ml in 1M HCl

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29031-19-4
Molecular Formula C6H15NO9S
Molecular Weight (g/mol) 277.244
MDL Number MFCD00080790
InChI Key FGNPLIQZJCYWLE-BTVCFUMJSA-N
Synonym d-glucosamine sulphate, 2r,3r,4s,5r-2-amino-3,4,5,6-tetrahydroxyhexanal sulfate, gevolox, glucosaminsulfat, sulfato de glucosamina, 2-amino-2-deoxy-d-glucose sulfate salt, glucosamine sulfate 2kcl, d-glucose, 2-amino-2-deoxy-, sulfate salt, glucosamine sulfate na salt, glucosamine sulfate dc
PubChem CID 115046
IUPAC Name (2R,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;sulfuric acid
SMILES C(C(C(C(C(C=O)N)O)O)O)O.OS(=O)(=O)O

Specifications

Color White
Quantity 25 g
Solubility Information Soluble at 1mg/ml in 1M HCl
Formula Weight 277.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Glucosamine sulfate
EINECSNumber 249-379-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.